5-Methyl-4-(3-methylbenzyl)-2,4-dihydro-3H-pyrazol-3-one ≥95%

Supplier: Matrix Scientific

047601-1G 047601-500MG
101880-736EA 181.54 USD
101880-736 101880-738
5-Methyl-4-(3-methylbenzyl)-2,4-dihydro-3H-pyrazol-3-one ≥95%
5-Methyl-4-(3-methylbenzyl)-2,4-dihydro-3H-pyrazol-3-one

Hazard: Irritant

Formula: C₁₂H₁₄N₂O
Melting Pt: 206…207 °C
MDL Number: MFCD11841006
CAS Number: 1173908-51-4

Order Now

Specification Test Results

Melting point 206-207°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR