1-(2,4-Difluorophenyl)-1H-pyrrole-2,5-dione ≥95%

Supplier: Matrix Scientific

047597-500MG 047597-1G
101880-720EA 181.54 USD
101880-720 101880-722
1-(2,4-Difluorophenyl)-1H-pyrrole-2,5-dione ≥95%
1-(2,4-Difluorophenyl)-1H-pyrrole-2,5-dione

Hazard: Irritant

Formula: C₁₀H₅F₂NO₂
Melting Pt: 83…85 °C
MDL Number: MFCD00033652
CAS Number: 6954-65-0

Order Now

Specification Test Results

Melting point 83-85°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR