2-[(4-Chlorophenyl)sulfanyl]acetohydrazide ≥95%

Supplier: Matrix Scientific

Synonyms: 2-[(4-Chlorophenyl)thio]acetohydrazide

047594-500MG 047594-1G
101880-710EA 181.54 USD
101880-710 101880-712
2-[(4-Chlorophenyl)sulfanyl]acetohydrazide ≥95%
2-[(4-Chlorophenyl)sulfanyl]acetohydrazide

Hazard: Irritant

Formula: C₈H₉ClN₂OS
MW: 216.69 g/mol
Melting Pt: 103…104 °C
MDL Number: MFCD01176459
CAS Number: 75150-40-2

Order Now

Specification Test Results

Melting point 103-104°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR