2-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Methyl-3-oxo-3,4-dihydro-2H-benzo[b][1,4]oxazine-6-carboxylic acid
047560-1G
047560-500MG
101880-596EA
927.69
USD
101880-596
101880-594
2-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylic acid ≥95%
2-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylic acid
Hazard: Irritant
Formula:
C₁₀H₉NO₄ MW: 207.19 g/mol Melting Pt: 263…265 °C |
MDL Number:
MFCD11207468 CAS Number: 1092352-65-2 |
Specification Test Results
Melting point | 263-265°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...