1-(4-Pyridinyl)-1-ethanone oxime ≥95%

Supplier: Matrix Scientific

047535-500MG
101880-476EA 181.54 USD
101880-476
1-(4-Pyridinyl)-1-ethanone oxime ≥95%
1-(4-Pyridinyl)-1-ethanone oxime

Hazard: Irritant

Formula: C₇H₈N₂O
Melting Pt: 156…157 °C
MDL Number: MFCD00185708
CAS Number: 1194-99-6

Order Now

Specification Test Results

Melting point 156-157°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR