1-(3-Pyridinyl)-1-ethanone oxime ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(Pyridin-3-yl)ethanone oxime

047534-500MG
101880-472EA 181.54 USD
101880-472
1-(3-Pyridinyl)-1-ethanone oxime ≥95%
1-(3-Pyridinyl)-1-ethanone oxime

Hazard: Irritant

Formula: C₇H₈N₂O
MW: 136.15 g/mol
Melting Pt: 106…107 °C
MDL Number: MFCD00179685
CAS Number: 5973-83-1

Order Now

Specification Test Results

Melting point 106-107°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR