1-(5-Chloro-2-nitrophenyl)-4-piperidinecarboxylicacid ≥95%

Supplier: Matrix Scientific

047530-1G
101880-464EA 467.69 USD
101880-464
1-(5-Chloro-2-nitrophenyl)-4-piperidinecarboxylicacid ≥95%
1-(5-Chloro-2-nitrophenyl)-4-piperidinecarboxylicacid

Hazard: Irritant

Formula: C₁₂H₁₃ClN₂O₄
Melting Pt: 103…104 °C
MDL Number: MFCD11553033
CAS Number: 847408-08-6

Order Now

Specification Test Results

Melting point 103-104°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR