4-Cyano-N-methoxy-N-methylbenzenecarboxamide ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Cyano-N-methoxy-N-methylbenzamide
047508-1G
101880-374EA
307.69
USD
101880-374
4-Cyano-N-methoxy-N-methylbenzenecarboxamide ≥95%
4-Cyano-N-methoxy-N-methylbenzenecarboxamide
Hazard: Irritant
Formula:
C₁₀H₁₀N₂O₂ MW: 190.2 g/mol Melting Pt: 78…80 °C |
MDL Number:
MFCD07367975 CAS Number: 116332-64-0 |
Specification Test Results
Melting point | 78-80°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...