Methyl-4-{2-[4-(benzyloxy)phenoxy]-ethoxy}benzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-{2-[4-(benzyloxy)phenoxy]ethoxy}benzoate

047505-1G
101880-358EA 350.77 USD
101880-358
Methyl-4-{2-[4-(benzyloxy)phenoxy]-ethoxy}benzenecarboxylate ≥95%
Methyl-4-{2-[4-(benzyloxy)phenoxy]-ethoxy}benzenecarboxylate

Hazard: Irritant

Formula: C₂₃H₂₂O₅
MW: 378.43 g/mol
Melting Pt: 163…165 °C
MDL Number: MFCD08689801
CAS Number: 937602-33-0

Order Now

Specification Test Results

Melting point 163-165°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR