4-(2,1,3-Benzothiadiazol-5-ylmethoxy)benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

047492-1G
101880-282EA 698.46 USD
101880-282
4-(2,1,3-Benzothiadiazol-5-ylmethoxy)benzenecarbaldehyde ≥95%
4-(2,1,3-Benzothiadiazol-5-ylmethoxy)benzenecarbaldehyde

Hazard: Irritant

Formula: C₁₄H₁₀N₂O₂S
MW: 270.31 g/mol
Melting Pt: 114…116 °C
MDL Number: MFCD08689796
CAS Number: 937602-23-8

Order Now

Specification Test Results

Melting point 114-116°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR