Methyl-4-(2,1,3-benzothiadiazol-5-ylmethoxy)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

047491-500MG 047491-5G
101880-274EA 556.92 USD
101880-274 101880-278
Methyl-4-(2,1,3-benzothiadiazol-5-ylmethoxy)benzenecarboxylate ≥95%
Methyl-4-(2,1,3-benzothiadiazol-5-ylmethoxy)benzenecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₂N₂O₃S
MW: 300.34 g/mol
Melting Pt: 135…137 °C
MDL Number: MFCD06808681
CAS Number: 874834-02-3

Order Now

Specification Test Results

Melting point 135-137°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR