Ethyl-4-(4-isopropylphenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%

Supplier: Matrix Scientific

047482-1G
101880-226EA 232.31 USD
101880-226
Ethyl-4-(4-isopropylphenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%
Ethyl-4-(4-isopropylphenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate

Hazard: Irritant

Formula: C₁₈H₂₄N₂O₃
Melting Pt: 140…142 °C
MDL Number: MFCD10179102
CAS Number: 219814-72-9

Order Now

Specification Test Results

Melting point 140-142°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR