Ethyl-4-(2-fluorophenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%

Supplier: Matrix Scientific

047480-1G
101880-214EA 232.31 USD
101880-214
Ethyl-4-(2-fluorophenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%
Ethyl-4-(2-fluorophenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₇FN₂O₃
Melting Pt: 157…159 °C
MDL Number: MFCD00548101
CAS Number: 313391-66-1

Order Now

Specification Test Results

Melting point 157-159°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR