4-(6-Bromo-2-pyridinyl)-1,3-thiazol-2-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-4-(6-bromopyridin-2-yl)-1,3-thiazole

047476-1G
101880-196EA 553.85 USD
101880-196
4-(6-Bromo-2-pyridinyl)-1,3-thiazol-2-amine ≥95%
4-(6-Bromo-2-pyridinyl)-1,3-thiazol-2-amine

Hazard: Irritant

Formula: C₈H₆BrN₃S
MW: 256.13 g/mol
Melting Pt: 245…246 °C
MDL Number: MFCD09027112
CAS Number: 937602-13-6

Order Now

Specification Test Results

Melting point 245-246°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR