5-Amino-1-(4-bromobenzyl)-1H-1,2,3-triazole-4-carboxamide ≥95%
Supplier: Matrix Scientific
047473-5G
047473-500MG
101880-180EA
556.92
USD
101880-180
101880-184
5-Amino-1-(4-bromobenzyl)-1H-1,2,3-triazole-4-carboxamide ≥95%
5-Amino-1-(4-bromobenzyl)-1H-1,2,3-triazole-4-carboxamide
Hazard: Irritant
Formula:
C₁₀H₁₀BrN₅O Melting Pt: 234 °C |
MDL Number:
MFCD07698597 CAS Number: 886362-30-7 |
Specification Test Results
Melting point | 234°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...