Ethyl-1-benzyl-5-methyl-1H-1,2,3-triazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

047468-1G
101880-152EA 467.69 USD
101880-152
Ethyl-1-benzyl-5-methyl-1H-1,2,3-triazole-4-carboxylate ≥95%
Ethyl-1-benzyl-5-methyl-1H-1,2,3-triazole-4-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₅N₃O₂
Melting Pt: 68 °C
MDL Number: MFCD06797393
CAS Number: 133992-58-2

Order Now

Specification Test Results

Melting point 68°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR