3-(Hydroxymethyl)-2(1H)-quinolinone ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(Hydroxymethyl)quinolin-2(1H)-one

047462-1G
101880-116EA 269.23 USD
101880-116
3-(Hydroxymethyl)-2(1H)-quinolinone ≥95%
3-(Hydroxymethyl)-2(1H)-quinolinone

Hazard: Irritant

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Melting Pt: 200 °C
MDL Number: MFCD06200995
CAS Number: 90097-45-3

Order Now

Specification Test Results

Melting point 200°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR