3-(4-Methylbenzyl)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-7-ol ≥95%

Supplier: Matrix Scientific

047454-1G
101880-072EA 698.46 USD
101880-072
3-(4-Methylbenzyl)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-7-ol ≥95%
3-(4-Methylbenzyl)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-7-ol

Hazard: Irritant

Formula: C₁₂H₁₁N₅O
Melting Pt: 203 °C
MDL Number: MFCD09909882
CAS Number: 258356-16-0

Order Now

Specification Test Results

Melting point 203°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR