1-[1-(4-Bromobenzyl)-5-methyl-1H-1,2,3-triazol-4-yl]-1-ethanol ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(4-Bromobenzyl)-4-(1-hydroxyethyl)-5-methyl-1H-1,2,3-triazole
047452-1G
101880-060EA
467.69
USD
101880-060
1-[1-(4-Bromobenzyl)-5-methyl-1H-1,2,3-triazol-4-yl]-1-ethanol ≥95%
1-[1-(4-Bromobenzyl)-5-methyl-1H-1,2,3-triazol-4-yl]-1-ethanol
Hazard: Irritant
Formula:
C₁₂H₁₄BrN₃O MW: 296.17 g/mol Melting Pt: 144 °C |
MDL Number:
MFCD09865012 CAS Number: 1017782-60-3 |
Specification Test Results
Melting point | 144°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...