1-(4-Bromobenzyl)-N-5-dimethyl-1H-1,2,3-triazole-4-carboxamide ≥95%
Supplier: Matrix Scientific
047451-500MG
047451-5G
101880-056EA
1073.85
USD
101880-056
101880-052
1-(4-Bromobenzyl)-N-5-dimethyl-1H-1,2,3-triazole-4-carboxamide ≥95%
1-(4-Bromobenzyl)-N-5-dimethyl-1H-1,2,3-triazole-4-carboxamide
Hazard: Irritant
Formula:
C₁₂H₁₃BrN₄O Melting Pt: 186 °C (dec.) |
MDL Number:
MFCD09607957 CAS Number: 952182-54-6 |
Specification Test Results
Melting point | 186°C(dec) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...