5-Methyl-1-(4-methylbenzyl)-1H-1,2,3-triazole-4-carboxylic acid ≥95%
Supplier: Matrix Scientific
047449-500MG
047449-5G
101880-044EA
1073.85
USD
101880-044
101880-040
5-Methyl-1-(4-methylbenzyl)-1H-1,2,3-triazole-4-carboxylic acid ≥95%
5-Methyl-1-(4-methylbenzyl)-1H-1,2,3-triazole-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₂H₁₃N₃O₂ Melting Pt: 199 °C |
MDL Number:
MFCD10758097 CAS Number: 1033463-40-9 |
Specification Test Results
Melting point | 199°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...