2-Oxo-1,2-dihydro-3-quinolinecarbonitrile ≥95%

Supplier: Matrix Scientific

047443-500MG 047443-5G
101880-004EA 418.46 USD
101880-004 101880-008
2-Oxo-1,2-dihydro-3-quinolinecarbonitrile ≥95%
2-Oxo-1,2-dihydro-3-quinolinecarbonitrile

Hazard: Irritant

Formula: C₁₀H₆N₂O
MW: 170.17 g/mol
Melting Pt: 314…318 °C
MDL Number: MFCD02175936
CAS Number: 36926-82-6

Order Now

Specification Test Results

Melting point 314-318°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR