3-(4-Bromobenzyl)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-7-ol ≥95%
Supplier: Matrix Scientific
047441-1G
101880-000EA
698.46
USD
101880-000
3-(4-Bromobenzyl)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-7-ol ≥95%
3-(4-Bromobenzyl)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-7-ol
Hazard: Irritant
Formula:
C₁₁H₈BrN₅O Melting Pt: >250 °C |
MDL Number:
MFCD09027111 CAS Number: 937604-16-5 |
Specification Test Results
Melting point |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...