4-Methoxy-2-(methylsulfanyl)-6-(trifluoromethyl)-5-pyrimidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

047425-500MG
101879-924EA 492.31 USD
101879-924
4-Methoxy-2-(methylsulfanyl)-6-(trifluoromethyl)-5-pyrimidinecarboxylic acid ≥95%
4-Methoxy-2-(methylsulfanyl)-6-(trifluoromethyl)-5-pyrimidinecarboxylic acid

Hazard: Irritant

Formula: C₈H₇F₃N₂O₃S
MW: 268.22 g/mol
Melting Pt: 158…159 °C
MDL Number: MFCD12827810
CAS Number: 1186404-89-6

Order Now

Specification Test Results

Melting point 158-159°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR