Bis(dicyclohexylamino)palladium acetate ≥95%

Supplier: Matrix Scientific

047417-500MG
101879-894EA 529.23 USD
101879-894
Bis(dicyclohexylamino)palladium acetate ≥95%
Bis(dicyclohexylamino)palladium acetate

Hazard: Irritant

Formula: C₂₈H₅₂N₂O₄Pd
Melting Pt: 140 °C (dec.)
MDL Number: MFCD09996893
CAS Number: 628339-96-8

Order Now

Specification Test Results

Melting point 140°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR