1-(tert-Butyl)-6-methyl-4-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-3-carbonitrile ≥95%

Supplier: Matrix Scientific

047413-1G
101879-874EA 387.69 USD
101879-874
1-(tert-Butyl)-6-methyl-4-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-3-carbonitrile ≥95%
1-(tert-Butyl)-6-methyl-4-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-3-carbonitrile

Hazard: Irritant

Formula: C₁₄H₁₄F₃N₃
Melting Pt: 162…164 °C
MDL Number: MFCD12827797
CAS Number: 1146221-74-0

Order Now

Specification Test Results

Melting point 162-164°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR