6-(Trifluoromethyl)-1H-indole-5-carbonitrile ≥95%
Supplier: Matrix Scientific
047409-1G
101879-858EA
592.31
USD
101879-858
6-(Trifluoromethyl)-1H-indole-5-carbonitrile ≥95%
6-(Trifluoromethyl)-1H-indole-5-carbonitrile
Hazard: Irritant
Formula:
C₁₀H₅F₃N₂ Melting Pt: 176 °C |
MDL Number:
MFCD12827791 CAS Number: 1186404-61-4 |
Specification Test Results
Melting point | 176°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...