3-(Benzyloxy)-4-nitrobenzenecarbohydrazide ≥95%

Supplier: Matrix Scientific

047406-500MG
101879-842EA 375.38 USD
101879-842
3-(Benzyloxy)-4-nitrobenzenecarbohydrazide ≥95%
3-(Benzyloxy)-4-nitrobenzenecarbohydrazide

Hazard: Irritant

Formula: C₁₄H₁₃N₃O₄
Melting Pt: 146…149 °C
MDL Number: MFCD12827788
CAS Number: 1186405-04-8

Order Now

Specification Test Results

Melting point 146-149°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR