Ethyl-3-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate ≥95%
Supplier: Matrix Scientific
047394-1G
101879-798EA
698.46
USD
101879-798
Ethyl-3-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate ≥95%
Ethyl 3-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Hazard: Irritant
Formula:
C₁₁H₈ClF₃N₂O₂ Melting Pt: 99…100 °C |
MDL Number:
MFCD11841001 CAS Number: 1135283-40-7 |
Specification Test Results
Melting point | 99-100°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...