Ethyl-8-iodo-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

047393-1G
101879-794EA 693.45 USD
101879-794
Ethyl-8-iodo-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate ≥95%
Ethyl 8-iodo-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₈F₃IN₂O₂
Melting Pt: 154…155 °C
MDL Number: MFCD11841000
CAS Number: 1135283-39-4

Order Now

Specification Test Results

Melting point 154-155°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR