5-Bromo-6-chloronicotinonitrile ≥95%
Supplier: Matrix Scientific
047390-500MG
101879-782EA
38.46
USD
101879-782
5-Bromo-6-chloronicotinonitrile ≥95%
5-Bromo-6-chloronicotinonitrile
Hazard: Irritant
Formula:
C₆H₂BrClN₂ Melting Pt: 156…158 °C |
MDL Number:
MFCD11840998 CAS Number: 71702-01-7 |
Specification Test Results
Melting point | 156-158°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...