4-Hydroxy-2-(2-pyridinyl)-5-pyrimidinecarboxylicacid ≥95%

Supplier: Matrix Scientific

047389-500MG
101879-778EA 233.85 USD
101879-778
4-Hydroxy-2-(2-pyridinyl)-5-pyrimidinecarboxylicacid ≥95%
4-Hydroxy-2-(2-pyridinyl)-5-pyrimidinecarboxylicacid

Hazard: Irritant

Formula: C₁₀H₇N₃O₃
MW: 217.19 g/mol
Melting Pt: 330 °C (dec.)
MDL Number: MFCD11100252
CAS Number: 56406-45-2

Order Now

Specification Test Results

Melting point 330°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR