6-Chloro-2,2'-bipyridine-5-carbonitrile ≥95%

Supplier: Matrix Scientific

047384-500MG
101879-758EA 556.92 USD
101879-758
6-Chloro-2,2'-bipyridine-5-carbonitrile ≥95%
6-Chloro-2,2'-bipyridine-5-carbonitrile

Hazard: Irritant

Formula: C₁₁H₆ClN₃
Melting Pt: 187…189 °C
MDL Number: MFCD09027897
CAS Number: 946385-03-1

Order Now

Specification Test Results

Melting point 187-189°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR