5-Chloro-6-hydroxynicotinonitrile ≥95%

Supplier: Matrix Scientific

047371-500MG
101879-702EA 500 USD
101879-702
5-Chloro-6-hydroxynicotinonitrile ≥95%
5-Chloro-6-hydroxynicotinonitrile

Hazard: Irritant

Formula: C₆H₃ClN₂O
Melting Pt: 242…244 °C
MDL Number: MFCD11100240
CAS Number: 19840-46-1

Order Now

Specification Test Results

Melting point 242-244°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR