6-Hydroxy-5-iodnicotinonitrile ≥95%
Supplier: Matrix Scientific
047370-500MG
101879-698EA
747.69
USD
101879-698
6-Hydroxy-5-iodnicotinonitrile ≥95%
6-Hydroxy-5-iodnicotinonitrile 2-(4-Formylphenyl)isonicotinonitrile
Hazard: Irritant
Formula:
C₆H₃IN₂O Melting Pt: 267…269 °C |
MDL Number:
MFCD11840996 CAS Number: 1135283-36-1 |
Specification Test Results
Melting point | 267-269°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...