1-(5-Chloro-2-pyridinyl)-3-(trifluoromethyl)-1H-pyrazol-5-ol ≥95%

Supplier: Matrix Scientific

047367-1G
101879-684EA 353.85 USD
101879-684
1-(5-Chloro-2-pyridinyl)-3-(trifluoromethyl)-1H-pyrazol-5-ol ≥95%
1-(5-Chloro-2-pyridinyl)-3-(trifluoromethyl)-1H-pyrazol-5-ol

Hazard: Irritant

Formula: C₉H₅ClF₃N₃O
Melting Pt: 60…62 °C
MDL Number: MFCD11100238
CAS Number: 189005-07-0

Order Now

Specification Test Results

Melting point 60-62°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR