8-Iodo-6-(trifluoromethyl)imidazo[1,2-a]pyridine ≥95%

Supplier: Matrix Scientific

047364-1G
101879-676EA 698.46 USD
101879-676
8-Iodo-6-(trifluoromethyl)imidazo[1,2-a]pyridine ≥95%
8-Iodo-6-(trifluoromethyl)imidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₈H₄F₃IN₂
Melting Pt: 114…116 °C
MDL Number: MFCD11100236
CAS Number: 1048913-78-5

Order Now

Specification Test Results

Melting point 114-116°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR