1-Boc-4-(aminomethyl)-4-hydroxypiperidine ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Boc-4-(Aminomethyl)-4-hydroxypiperidinetert-Butyl 4-(aminomethyl)-4-hydroxypiperidine-1-carboxylate

047363-1G
101879-672EA 138.46 USD
101879-672
1-Boc-4-(aminomethyl)-4-hydroxypiperidine ≥95%
1-Boc-4-(aminomethyl)-4-hydroxypiperidine

Hazard: Irritant

Formula: C₁₁H₂₂N₂O₃
MW: 230.3 g/mol
Melting Pt: >300 °C
MDL Number: MFCD11579803
CAS Number: 392331-66-7

Order Now

Specification Test Results

Melting point 300°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR