6-Chloro[1,2,4]triazolo[4,3-a]pyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
6-Chloro-[1,2,4]triazolo[4,3-a]pyridine
047362-1G
101879-668EA
483.08
USD
101879-668
6-Chloro[1,2,4]triazolo[4,3-a]pyridine ≥95%
6-Chloro[1,2,4]triazolo[4,3-a]pyridine
Hazard: Irritant
Formula:
C₆H₄ClN₃ MW: 153.57 g/mol Melting Pt: 141…143 °C |
MDL Number:
MFCD11100235 CAS Number: 1048913-70-7 |
Specification Test Results
Melting point | 141-143°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...