4-(Benzylamino)-6-chloro-5-pyrimidinecarbaldehyde ≥95%
Supplier: Matrix Scientific
047353-500MG
101879-632EA
375.38
USD
101879-632
4-(Benzylamino)-6-chloro-5-pyrimidinecarbaldehyde ≥95%
4-(Benzylamino)-6-chloro-5-pyrimidinecarbaldehyde
Hazard: Irritant
Formula:
C₁₂H₁₀ClN₃O Melting Pt: 70…72 °C |
MDL Number:
MFCD11840994 CAS Number: 59311-82-9 |
Specification Test Results
Melting point | 70-72°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...