5-(4-Aminophenyl)-1,3,4-oxadiazol-2-ol ≥95%

Supplier: Matrix Scientific

047352-500MG
101879-628EA 707.69 USD
101879-628
5-(4-Aminophenyl)-1,3,4-oxadiazol-2-ol ≥95%
5-(4-Aminophenyl)-1,3,4-oxadiazol-2-ol

Hazard: Irritant

Formula: C₈H₇N₃O₂
Melting Pt: 171…173 °C
MDL Number: MFCD02182673
CAS Number: 113118-47-1

Order Now

Specification Test Results

Melting point 171-173°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR