5-Amino-1-benzofuran-2-carbohydrazide ≥95%

Supplier: Matrix Scientific

047351-500MG
101879-624EA 747.69 USD
101879-624
5-Amino-1-benzofuran-2-carbohydrazide ≥95%
5-Amino-1-benzofuran-2-carbohydrazide

Hazard: Irritant

Formula: C₉H₉N₃O₂
Melting Pt: 181…183 °C
MDL Number: MFCD11100227
CAS Number: 1048913-38-7

Order Now

Specification Test Results

Melting point 181-183°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR