3-Amino-4-iodo-1H-indazole ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Iodo-1H-indazol-3-amine

047346-5G
102551-374EA 623.08 USD
102551-374
3-Amino-4-iodo-1H-indazole ≥95%
3-Amino-4-iodo-1H-indazole

Hazard: Irritant

Formula: C₇H₆IN₃
MW: 259.0471 g/mol
Melting Pt: 154…155 °C
MDL Number: MFCD03426697
CAS Number: 599191-73-8

Order Now

Specification Test Results

Melting point 154-155°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR