3-Oxo-3-(3-thienyl)propanenitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Oxo-3-(thiophen-3-yl)propanenitrile

047341-1G
101879-586EA 467.69 USD
101879-586
3-Oxo-3-(3-thienyl)propanenitrile ≥95%
3-Oxo-3-(3-thienyl)propanenitrile

Hazard: Irritant

Formula: C₇H₅NOS
MW: 151.19 g/mol
Melting Pt: 91…93 °C
MDL Number: MFCD00111016
CAS Number: 69879-30-7

Order Now

Specification Test Results

Melting point 91-93°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR