2-(Diphenylamino)-1,3-thiazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

047336-500MG
101879-566EA 258.46 USD
101879-566
2-(Diphenylamino)-1,3-thiazole-4-carboxylic acid ≥95%
2-(Diphenylamino)-1,3-thiazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₂N₂O₂S
Melting Pt: 222…224 °C
MDL Number: MFCD08689754
CAS Number: 937601-81-5

Order Now

Specification Test Results

Melting point 222-224°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR