1-(4-Bromophenyl)-2-(methylsulfonyl)-1-ethanone ≥95%
Supplier: Matrix Scientific
047333-1G
101879-550EA
350.77
USD
101879-550
1-(4-Bromophenyl)-2-(methylsulfonyl)-1-ethanone ≥95%
1-(4-Bromophenyl)-2-(methylsulfonyl)-1-ethanone
Hazard: Irritant
Formula:
C₉H₉BrO₃S MW: 277.14 g/mol Melting Pt: 152…154 °C |
MDL Number:
MFCD09864834 CAS Number: 500576-33-0 |
Specification Test Results
Melting point | 152-154°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...