2-Anilino-1,3-thiazole-4-carboxylic acid ≥95%
Supplier: Matrix Scientific
047328-500MG
101879-522EA
172.31
USD
101879-522
2-Anilino-1,3-thiazole-4-carboxylic acid ≥95%
2-Anilino-1,3-thiazole-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₀H₈N₂O₂S MW: 220.25 g/mol Melting Pt: 278…280 °C |
MDL Number:
MFCD04117336 CAS Number: 165683-01-2 |
Specification Test Results
Melting point | 278-280°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...