5-Amino-1-methyl-1H-pyrazole-4-carboxamide ≥95%
Supplier: Matrix Scientific
047326-1G
101879-514EA
133.85
USD
101879-514
5-Amino-1-methyl-1H-pyrazole-4-carboxamide ≥95%
5-Amino-1-methyl-1H-pyrazole-4-carboxamide
Hazard: Irritant
Formula:
C₅H₈N₄O MW: 140.15 g/mol Melting Pt: 223…235 °C |
MDL Number:
MFCD00111808 CAS Number: 18213-75-7 |
Specification Test Results
Melting point | 223-235°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...