4-(Trifluoromethyl)-1-benzothiophene-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Trifluoromethyl)benzo[b]thiophene-2-carboxylic acid

047322-1G
101879-496EA 467.69 USD
101879-496
4-(Trifluoromethyl)-1-benzothiophene-2-carboxylic acid ≥95%
4-(Trifluoromethyl)-1-benzothiophene-2-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₅F₃O₂S
MW: 246.21 g/mol
Melting Pt: 188…190 °C
MDL Number: MFCD09027109
CAS Number: 826995-55-5

Order Now

Specification Test Results

Melting point 188-190°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR