6-Chloro-1-benzothiophene-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Chlorobenzo[b]thiophene-2-carboxylic acid

047320-1G
101879-486EA 60 USD
101879-486
6-Chloro-1-benzothiophene-2-carboxylic acid ≥95%
6-Chloro-1-benzothiophene-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₅ClO₂S
MW: 212.66 g/mol
Melting Pt: 271…274 °C
MDL Number: MFCD07377059
CAS Number: 26018-73-5

Order Now

Specification Test Results

Melting point 271-274°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR